Assay Details
| Gene Symbol: Mettl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029790.3 | NP_084066.2 | 3 - 4 | 520 | 75 | |
| GenBank mRNA | AK002682.1 | - | 3 - 4 | 521 | 75 | |
| BC089533.1 | - | 4 - 5 | 647 | 75 | ||
| BC106186.1 | - | 3 - 4 | 492 | 30244889 | 75 | |
| BC115926.1 | - | 3 - 4 | 479 | 40105262 | 75 | |
| BC117549.1 | - | 3 - 4 | 479 | 40105259 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl15
Genomic Map