Assay Details
Gene Symbol: Fam20c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030565.6 | NP_085042.2 | 8 - 9 | 1980 | 72 | |
GenBank mRNA | AK149780.1 | - | 9 - 10 | 2060 | 72 | |
AK154810.1 | - | 8 - 9 | 1976 | 72 | ||
AK220286.1 | - | 8 - 9 | 1496 | 72 | ||
AY778962.1 | - | 8 - 9 | 1961 | 72 | ||
BC004044.1 | - | 5 - 6 | 589 | 72 | ||
BC025814.1 | - | 8 - 9 | 1931 | 72 | ||
BC025826.1 | - | 8 - 9 | 1961 | 72 | ||
CT010363.1 | - | 4 - 5 | 435 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
