Assay Details
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030697.2 | NP_109622.1 | 9 - 10 | 2304 | 74 | |
GenBank mRNA | AK002623.1 | - | 9 - 10 | 2305 | 74 | |
AK132011.1 | - | 9 - 10 | 2326 | 74 | ||
BC037741.1 | - | 4 - 5 | 438 | 74 | ||
BC061055.1 | - | 4 - 5 | 450 | 74 | ||
BC115933.1 | - | 3 - 4 | 408 | 74 | ||
KJ650203.1 | - | 8 - 9 | 2235 | 74 | ||
KJ663713.1 | - | 8 - 9 | 2076 | 74 | ||
KJ663714.1 | - | 9 - 10 | 2097 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
