Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900010299.mm.1 |
NM_053159.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010299.mm.1 |
NM_053159.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010299.mm.1 |
TC0900001318.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010300.mm.1 |
NM_053159.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010300.mm.1 |
NM_053159.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010300.mm.1 |
TC0900001318.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010298.mm.1 |
NM_053159.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010298.mm.1 |
NM_053159.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010298.mm.1 |
TC0900001318.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010301.mm.1 |
NM_053159.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010301.mm.1 |
NM_053159.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010301.mm.1 |
TC0900001318.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010295.mm.1 |
NM_053159.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010295.mm.1 |
NM_053159.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010295.mm.1 |
TC0900001318.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010302.mm.1 |
NM_053159.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010302.mm.1 |
NM_053159.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010302.mm.1 |
TC0900001318.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010303.mm.1 |
NM_053159.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010303.mm.1 |
NM_053159.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010303.mm.1 |
TC0900001318.mm.1 |
MTA TC |