Assay Details
| Gene Symbol: Hsd17b11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053262.3 | NP_444492.1 | 1 - 2 | 268 | 94 | |
| GenBank mRNA | AF304306.1 | - | 1 - 2 | 264 | 94 | |
| AK049355.1 | - | 1 - 2 | 312 | 94 | ||
| AK154803.1 | - | 1 - 2 | 283 | 94 | ||
| AK155174.1 | - | 1 - 2 | 283 | 94 | ||
| AK155202.1 | - | 1 - 2 | 288 | 94 | ||
| AK155327.1 | - | 1 - 2 | 288 | 94 | ||
| AK158327.1 | - | 1 - 2 | 283 | 94 | ||
| AK170939.1 | - | 1 - 2 | 283 | 94 | ||
| AY053570.1 | - | 1 - 2 | 282 | 94 | ||
| BC038340.1 | - | 1 - 2 | 268 | 4458725 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b11
Genomic Map