Assay Details
Gene Symbol: B3galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080445.4 | NP_536693.1 | 1 | 508 | 61 | |
GenBank mRNA | AK033608.1 | - | 1 | 418 | 61 | |
AK078083.1 | - | 1 | 480 | 61 | ||
AK090344.1 | - | 1 | 450 | 61 | ||
AY050569.1 | - | 1 | 452 | 61 | ||
BC057933.1 | - | 1 | 460 | 61 | ||
BC082998.1 | - | 1 | 455 | 5011578 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycosaminoglycan biosynthetic process
protein glycosylation
galactosyltransferase activity
UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
UDP-galactosyltransferase activity
galactosylxylosylprotein 3-beta-galactosyltransferase activity
glycosaminoglycan biosynthetic process
protein glycosylation
galactosyltransferase activity
UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
UDP-galactosyltransferase activity
galactosylxylosylprotein 3-beta-galactosyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441753 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441749 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441755 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441750 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441754 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441751 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441752 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2467415 | AK090344.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148617 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52182 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285063 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54529 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140311 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140452 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162682 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2467415 | AK090344.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P211334 | BC057933.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P211334 | AY050569.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P211334 | BC082998.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P211334 | AK078083.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P211334 | AK090344.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441725 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441739 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441728 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441742 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140311 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140452 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162682 | - | - |
Whole Mouse Genome G4122A | A_51_P211334 | BC057933.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P211334 | AY050569.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P211334 | BC082998.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P211334 | AK078083.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P211334 | AK090344.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124712 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162682 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140311 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140452 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03673258 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03673261 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441749 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441750 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441753 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441752 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441754 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441751 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03477240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03478522 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03412714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04055152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03412713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03478521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441739 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10519236 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519238 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519235 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519237 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1421155_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435252_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1440340_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10519234 | - | - |
Mouse Genome 430 2.0 Array | 1421155_at | - | - |
Mouse Genome 430 2.0 Array | 1435252_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440340_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1421155_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440340_at | - | - |
Mouse Genome 430 2.0 Array | 1435252_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 117592_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 117592_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400034965.mm.1 | TC0400004163.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400034965.mm.1 | NM_080445.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034965.mm.1 | NM_080445.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034964.mm.1 | TC0400004163.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400034964.mm.1 | NM_080445.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034964.mm.1 | NM_080445.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034967.mm.1 | TC0400004163.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400034967.mm.1 | NM_080445.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034967.mm.1 | NM_080445.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034963.mm.1 | TC0400004163.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400034963.mm.1 | NM_080445.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034963.mm.1 | NM_080445.4 | Refseq |
Mouse Expression Set 430 | 1435252_PM_at | - | - |
Mouse Expression Set 430 | 1435252_at | - | - |
Mouse Expression Set 430 | 1421155_PM_at | - | - |
Mouse Expression Set 430 | 1421155_at | - | - |
Mouse Expression Set 430 | 1440340_PM_at | - | - |
Mouse Expression Set 430 | 1440340_at | - | - |
Murine Genome U74v2 Set | 98291_PM_at | - | - |
Murine Genome U74v2 Set | 98291_at | - | - |
Murine Genome U74v2 Set | 166424_at | - | - |
Murine Genome U74v2 Set | 166424_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1435252_at | - | - |
Mouse Genome 430A 2.0 Array | 1421155_at | - | - |
Mouse Genome 430A 2.0 Array | 1435252_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1421155_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 606366 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 104060519 | - | - |
MouseWG-6 v2 Expression BeadChip | 50195 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 50195 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 104060519 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000050195 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2687140 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2687140 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2488783 | - | - |
mouse-6 v1.1 (Illumina) | scl00117592.2_262-S | - | - |
mouse-6 v1.1 (Illumina) | scl23423.1_248-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 104060519 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 50195 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00117592.2_262-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104060519 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 50195 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2687140 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2687140 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2488783 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00117592.2_262-S | - | - |
