Assay Details
| Gene Symbol: Helb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080446.2 | NP_536694.2 | 3 - 4 | 845 | 65 | |
| GenBank mRNA | AB048542.1 | - | 3 - 4 | 770 | 65 | |
| AK131122.1 | - | 3 - 4 | 867 | 65 | ||
| BC068140.1 | - | 3 - 4 | 773 | 6843107 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Helb
Genomic Map