Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_001130163.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_001130163.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_001130164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_001130164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_001130165.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_001130165.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_001130166.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_001130166.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_130885.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
NM_130885.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002339.mm.1 |
TC1500000331.mm.1 |
MTA TC |