Assay Details
| Gene Symbol: Arr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133205.3 | NP_573468.1 | 11 - 12 | 872 | 105 | |
| GenBank mRNA | AF156979.1 | - | 11 - 12 | 813 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arr3
Genomic Map