Assay Details
| Gene Symbol: Rbm39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291114.1 | NP_001278043.1 | 1 - 2 | 354 | 135 | |
| NM_133242.3 | NP_573505.2 | 1 - 2 | 354 | 135 | ||
| GenBank mRNA | AK005227.1 | - | 1 - 2 | 302 | 135 | |
| AK005234.1 | - | 1 - 2 | 303 | 135 | ||
| AK009719.1 | - | 1 - 2 | 267 | 135 | ||
| AK147076.1 | - | 1 - 2 | 299 | 135 | ||
| AK154989.1 | - | 1 - 2 | 285 | 135 | ||
| AK156737.1 | - | 1 - 2 | 263 | 135 | ||
| AK167355.1 | - | 1 - 2 | 299 | 135 | ||
| BC023205.1 | - | 1 - 2 | 256 | 135 | ||
| BC086645.1 | - | 1 - 2 | 277 | 6817169 | 135 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK147076.1,AK154989.1,AK167355.1,BC086645.1,NM_001291114.1,NM_133242.3 transcripts.
Set Membership: |
|

Rbm39
Genomic Map