Assay Details
| Gene Symbol: Rbm39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291114.1 | NP_001278043.1 | 5 - 6 | 726 | 83 | |
| NM_001291115.1 | NP_001278044.1 | 5 - 6 | 521 | 83 | ||
| NM_133242.3 | NP_573505.2 | 5 - 6 | 726 | 83 | ||
| GenBank mRNA | AK081994.1 | - | 1 - 2 | 463 | 83 | |
| AK135289.1 | - | 4 - 5 | 352 | 83 | ||
| AK146046.1 | - | 5 - 6 | 521 | 83 | ||
| AK147076.1 | - | 5 - 6 | 671 | 83 | ||
| AK154989.1 | - | 5 - 6 | 657 | 83 | ||
| BC004000.1 | - | 1 - 2 | 58 | 83 | ||
| BC023205.1 | - | 5 - 6 | 628 | 83 | ||
| BC030493.1 | - | 3 - 4 | 341 | 5400675 | 83 | |
| BC082607.1 | - | 3 - 4 | 316 | 30667785 | 83 | |
| BC086645.1 | - | 5 - 6 | 649 | 6817169 | 83 | |
| BC092268.1 | - | 3 - 4 | 475 | 83 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC082607.1 transcripts.
Set Membership: |
|

Rbm39
Genomic Map