Assay Details
| Gene Symbol: Mutyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159581.1 | NP_001153053.1 | 9 - 10 | 737 | 69 | |
| NM_133250.2 | NP_573513.2 | 8 - 9 | 644 | 69 | ||
| GenBank mRNA | AB117937.1 | - | 6 - 7 | 532 | 69 | |
| AB117938.1 | - | 7 - 8 | 606 | 69 | ||
| AK077546.1 | - | 8 - 9 | 641 | 69 | ||
| AK156790.1 | - | 9 - 10 | 727 | 69 | ||
| AY007717.1 | - | 7 - 8 | 600 | 69 | ||
| BC057942.1 | - | 8 - 9 | 625 | 5320744 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mutyh
Genomic Map