Assay Details
| Gene Symbol: Grid2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159321.1 | NP_001152793.1 | 7 - 8 | 1450 | 84 | |
| NM_133355.1 | NP_579933.1 | 6 - 7 | 754 | 84 | ||
| GenBank mRNA | AF099933.2 | - | 6 - 7 | 754 | 84 | |
| BC075624.1 | - | 806 | 6841572 | 84 | ||
| DQ193535.1 | - | 7 - 8 | 1450 | 84 | ||
More Information
Set Membership: |
|

Grid2ip
Genomic Map