Assay Details
| Gene Symbol: Ces1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133660.3 | NP_598421.1 | 3 - 4 | 599 | 72 | |
| GenBank mRNA | AK144549.1 | - | 3 - 4 | 584 | 72 | |
| BC019208.1 | - | 3 - 4 | 530 | 5052893 | 72 | |
| S80191.1 | - | 3 - 4 | 529 | 72 | ||
More Information
Set Membership: |
|

Ces1e
Genomic Map