Assay Details
| Gene Symbol: Ndufv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133666.3 | NP_598427.1 | 6 - 7 | 1047 | 56 | |
| GenBank mRNA | AK005021.1 | - | 6 - 7 | 995 | 56 | |
| AK075692.1 | - | 6 - 7 | 992 | 56 | ||
| AK151455.1 | - | 6 - 7 | 958 | 56 | ||
| AK178320.1 | - | 2 - 3 | 235 | 56 | ||
| AK188080.1 | - | 1 - 2 | 130 | 56 | ||
| AK188929.1 | - | 1 - 2 | 117 | 56 | ||
| AK205680.1 | - | 1 - 2 | 108 | 56 | ||
| AK209435.1 | - | 2 - 3 | 235 | 56 | ||
| AK210493.1 | - | 2 - 3 | 235 | 56 | ||
| AK218408.1 | - | 2 - 3 | 234 | 56 | ||
| BC014818.1 | - | 6 - 7 | 1017 | 4456931 | 56 | |
| BC041682.1 | - | 6 - 7 | 950 | 4221001 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufv1
Genomic Map