Assay Details
| Gene Symbol: Ppp1r32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133689.1 | NP_598450.1 | 9 - 10 | 927 | 99 | |
| GenBank mRNA | AK005821.1 | - | 9 - 10 | 960 | 99 | |
| AK016325.1 | - | 9 - 10 | 955 | 99 | ||
| AY032666.1 | - | 9 - 10 | 927 | 99 | ||
| BC118523.1 | - | 9 - 10 | 903 | 40105249 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r32
Genomic Map