Assay Details
Gene Symbol: Ppp1r32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133689.1 | NP_598450.1 | 10 - 11 | 1050 | 98 | |
GenBank mRNA | AK005821.1 | - | 10 - 11 | 1083 | 98 | |
AK016325.1 | - | 10 - 11 | 1078 | 98 | ||
AY032666.1 | - | 10 - 11 | 1050 | 98 | ||
BC118523.1 | - | 10 - 11 | 1026 | 40105249 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
