Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200010339.mm.1 |
NM_001159632.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010339.mm.1 |
NM_001159632.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010339.mm.1 |
NM_133699.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010339.mm.1 |
NM_133699.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010339.mm.1 |
TC1200001468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200010334.mm.1 |
NM_001159632.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010334.mm.1 |
NM_001159632.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010334.mm.1 |
TC1200001468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200010336.mm.1 |
NM_001159632.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010336.mm.1 |
NM_001159632.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010336.mm.1 |
NM_133699.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010336.mm.1 |
NM_133699.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010336.mm.1 |
TC1200001468.mm.1 |
MTA TC |