Assay Details
| Gene Symbol: Kptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133727.2 | NP_598488.2 | 8 - 9 | 871 | 76 | |
| GenBank mRNA | AK009746.1 | - | 8 - 9 | 852 | 76 | |
| BC018316.1 | - | 8 - 9 | 839 | 4160465 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kptn
Genomic Map