Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500032938.mm.1 |
TC0500003590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032938.mm.1 |
NM_133735.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032938.mm.1 |
NM_133735.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032941.mm.1 |
TC0500003590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032941.mm.1 |
NM_133735.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032941.mm.1 |
NM_133735.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032942.mm.1 |
TC0500003590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032942.mm.1 |
NM_133735.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032942.mm.1 |
NM_133735.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032943.mm.1 |
TC0500003590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032943.mm.1 |
NM_133735.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032943.mm.1 |
NM_133735.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032937.mm.1 |
TC0500003590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032937.mm.1 |
NM_133735.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032937.mm.1 |
NM_133735.2 |
Refseq |