Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900013724.mm.1 |
NM_001164572.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013724.mm.1 |
NM_001164572.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013724.mm.1 |
NM_133741.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013724.mm.1 |
NM_133741.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013724.mm.1 |
TC0900001635.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013723.mm.1 |
NM_001164572.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013723.mm.1 |
NM_001164572.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013723.mm.1 |
NM_133741.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013723.mm.1 |
NM_133741.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013723.mm.1 |
TC0900001635.mm.1 |
MTA TC |