Assay Details
| Gene Symbol: Ssbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133772.2 | NP_598533.2 | 9 - 10 | 778 | 83 | |
| GenBank mRNA | AK004835.1 | - | 9 - 10 | 891 | 83 | |
| AK154336.1 | - | 9 - 10 | 779 | 83 | ||
| BC002150.1 | - | 9 - 10 | 737 | 3484896 | 83 | |
| CX237617.1 | - | 8 - 9 | 560 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssbp4
Genomic Map