Assay Details
Gene Symbol: Ssbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133772.2 | NP_598533.2 | 15 - 16 | 1091 | 99 | |
GenBank mRNA | AK004835.1 | - | 15 - 16 | 1204 | 99 | |
AK154336.1 | - | 15 - 16 | 1092 | 99 | ||
AK157387.1 | - | 14 - 15 | 1171 | 99 | ||
BC002150.1 | - | 15 - 16 | 1050 | 3484896 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
