Assay Details
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133803.2 | NP_598564.2 | 3 - 4 | 434 | 85 | |
GenBank mRNA | AK031020.1 | - | 3 - 4 | 399 | 85 | |
AK039975.1 | - | 3 - 4 | 395 | 85 | ||
AK087994.1 | - | 3 - 4 | 398 | 85 | ||
AK149853.1 | - | 3 - 4 | 399 | 85 | ||
AK150101.1 | - | 3 - 4 | 394 | 85 | ||
AK150126.1 | - | 3 - 4 | 388 | 85 | ||
AK168195.1 | - | 3 - 4 | 387 | 85 | ||
BC004600.1 | - | 2 - 3 | 376 | 3584924 | 85 | |
BC066817.1 | - | 3 - 4 | 385 | 30429618 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
