Assay Details
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133803.2 | NP_598564.2 | 9 - 10 | 1063 | 73 | |
GenBank mRNA | AK031020.1 | - | 9 - 10 | 1028 | 73 | |
AK039975.1 | - | 9 - 10 | 1024 | 73 | ||
AK087994.1 | - | 9 - 10 | 1027 | 73 | ||
AK149853.1 | - | 9 - 10 | 1028 | 73 | ||
AK150101.1 | - | 9 - 10 | 1023 | 73 | ||
AK150126.1 | - | 9 - 10 | 1017 | 73 | ||
AK166842.1 | - | 9 - 10 | 1028 | 73 | ||
AK168195.1 | - | 9 - 10 | 1016 | 73 | ||
AK207387.1 | - | 2 - 3 | 99 | 73 | ||
AK208198.1 | - | 2 - 3 | 99 | 73 | ||
BC004600.1 | - | 8 - 9 | 1005 | 3584924 | 73 | |
BC066817.1 | - | 9 - 10 | 1014 | 30429618 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
