Assay Details
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133803.2 | NP_598564.2 | 14 - 15 | 1650 | 63 | |
GenBank mRNA | AK031020.1 | - | 14 - 15 | 1615 | 63 | |
AK087994.1 | - | 14 - 15 | 1614 | 63 | ||
AK149853.1 | - | 14 - 15 | 1615 | 63 | ||
AK150101.1 | - | 14 - 15 | 1610 | 63 | ||
AK150126.1 | - | 14 - 15 | 1604 | 63 | ||
AK166842.1 | - | 14 - 15 | 1615 | 63 | ||
AK168195.1 | - | 14 - 15 | 1603 | 63 | ||
AK183507.1 | - | 1 - 2 | 50 | 63 | ||
AK184883.1 | - | 1 - 2 | 97 | 63 | ||
AK209237.1 | - | 1 - 2 | 50 | 63 | ||
AK218877.1 | - | 1 - 2 | 50 | 63 | ||
BC004600.1 | - | 13 - 14 | 1592 | 3584924 | 63 | |
BC066817.1 | - | 14 - 15 | 1601 | 30429618 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
