Assay Details
| Gene Symbol: Cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133837.4 | NP_598598.1 | 8 - 9 | 617 | 120 | |
| GenBank mRNA | AK090122.1 | - | 8 - 9 | 618 | 120 | |
| AK141463.1 | - | 8 - 9 | 605 | 120 | ||
| AK159208.1 | - | 8 - 9 | 638 | 120 | ||
| BC019753.1 | - | 8 - 9 | 592 | 5008314 | 120 | |
| BC022972.1 | - | 8 - 9 | 606 | 5368775 | 120 | |
| BC024787.1 | - | 8 - 9 | 604 | 5359215 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc123
Genomic Map