Assay Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133847.3 | NP_598608.2 | 17 - 18 | 2045 | 73 | |
| GenBank mRNA | AB093220.2 | - | 16 - 17 | 1789 | 73 | |
| AK048158.1 | - | 17 - 18 | 1803 | 73 | ||
| AK083541.1 | - | 17 - 18 | 2045 | 73 | ||
| AK087048.1 | - | 17 - 18 | 1798 | 73 | ||
| AK088012.1 | - | 17 - 18 | 1801 | 73 | ||
| AK162435.1 | - | 17 - 18 | 1806 | 73 | ||
| AK193992.1 | - | 1 - 2 | 44 | 73 | ||
| AK201108.1 | - | 1 - 2 | 44 | 73 | ||
| AK218670.1 | - | 1 - 2 | 44 | 73 | ||
| BC006741.1 | - | 3 - 4 | 482 | 73 | ||
| BC071208.1 | - | 4 - 5 | 864 | 73 | ||
| BC110309.1 | - | 17 - 18 | 1801 | 5324218 | 73 | |
| BC116320.2 | - | 17 - 18 | 1997 | 40096002 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | BC110309.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | BC006741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | BC071208.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | AK171487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | AK162435.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | BC110309.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | BC006741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | BC071208.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | AK171487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P483922 | AK162435.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323890 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392929 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213662 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646309 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659096 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783412 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 217905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756461 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922304 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140253 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713958 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1735 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81627 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | BC110309.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | BC006741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | BC071208.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | AK171487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | AK162435.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | BC110309.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | BC006741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | BC071208.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | AK171487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P483922 | AK162435.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P198993 | BC110309.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P198993 | BC116320.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P198993 | AK210804.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P198993 | AK171487.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P198993 | AK162435.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | BC110309.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | BC006741.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | BC071208.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | AK171487.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | AK162435.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | BC110309.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | BC006741.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | BC071208.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | AK171487.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483922 | AK162435.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753257 | - | - |
| Whole Mouse Genome G4122A | A_52_P198993 | BC110309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198993 | BC116320.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198993 | AK210804.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198993 | AK171487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198993 | AK162435.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | BC110309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | BC006741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | BC071208.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | AK171487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | AK162435.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | BC110309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | BC006741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | BC071208.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | AK171487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483922 | AK162435.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127777 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105446 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02245159 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02245169 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01415154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02245162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21753257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01649720 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10477281 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477278 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477275 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477266 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477267 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477274 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477283 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477276 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477272 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477268 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477282 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477277 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477284 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477265 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477280 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477279 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477270 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477273 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477285 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423204_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423205_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10477264 | - | - |
| Mouse Genome 430 2.0 Array | 1423204_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423205_at | - | - |
| Mouse Genome 430 2.0 Array | 1423204_at | - | - |
| Mouse Genome 430 2.0 Array | 1423205_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 99237_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 99237_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200018153.mm.1 | NM_133847.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018153.mm.1 | NM_133847.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018153.mm.1 | TC0200002310.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200018152.mm.1 | NM_133847.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018152.mm.1 | NM_133847.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018152.mm.1 | TC0200002310.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423205_at | - | - |
| Mouse Expression Set 430 | 1423204_at | - | - |
| Mouse Expression Set 430 | 1423204_PM_at | - | - |
| Mouse Expression Set 430 | 1423205_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423204_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423205_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423204_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423205_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423204_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423204_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423205_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423205_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 344178 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 450538 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 450538 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1050093 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4210112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004210112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050093 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000450538 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220640 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2501619 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247441 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220640 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247441 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2501619 | - | - |
| mouse-6 v1.1 (Illumina) | scl20100.20_8-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003215.1_26-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0099237.1_73-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050093 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4210112 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 450538 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0099237.1_73-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003215.1_26-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20100.20_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 450538 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050093 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2977690 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220640 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220640 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2977690 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20100.20_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0099237.1_73-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003215.1_26-S | - | - |

Tm9sf4
Genomic Map