Assay Details
| Gene Symbol: Ttll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142732.1 | NP_001136204.1 | 2 - 3 | 377 | 66 | |
| NM_133923.6 | NP_598684.4 | 2 - 3 | 494 | 66 | ||
| GenBank mRNA | AK029462.1 | - | 2 - 3 | 378 | 66 | |
| AM690746.1 | - | 2 - 3 | 494 | 66 | ||
| BC021404.1 | - | 1 - 2 | 126 | 66 | ||
More Information
Set Membership: |
|

Ttll3
Genomic Map