Assay Details
| Gene Symbol: Itfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133927.1 | NP_598688.1 | 3 - 4 | 336 | 81 | |
| GenBank mRNA | AK134630.1 | - | 3 - 4 | 357 | 81 | |
| AK147728.1 | - | 3 - 4 | 338 | 81 | ||
| AK159637.1 | - | 3 - 4 | 345 | 81 | ||
| BC014833.1 | - | 3 - 4 | 336 | 4507661 | 81 | |
| BC025492.1 | - | 3 - 4 | 356 | 5309950 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itfg2
Genomic Map