Assay Details
| Gene Symbol: Itfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133927.1 | NP_598688.1 | 8 - 9 | 943 | 71 | |
| GenBank mRNA | AK134630.1 | - | 8 - 9 | 964 | 71 | |
| AK147728.1 | - | 8 - 9 | 945 | 71 | ||
| AK159637.1 | - | 8 - 9 | 952 | 71 | ||
| BC014833.1 | - | 8 - 9 | 943 | 4507661 | 71 | |
| BC025492.1 | - | 8 - 9 | 963 | 5309950 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itfg2
Genomic Map