Assay Details
| Gene Symbol: Rpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133933.4 | NP_598694.3 | 4 - 5 | 909 | 81 | |
| GenBank mRNA | AK029923.1 | - | 4 - 5 | 867 | 81 | |
| AK052130.1 | - | 4 - 5 | 863 | 81 | ||
| AK132750.1 | - | 4 - 5 | 882 | 81 | ||
| AK151899.1 | - | 4 - 5 | 873 | 81 | ||
| AK152003.1 | - | 4 - 5 | 882 | 81 | ||
| AK153867.1 | - | 4 - 5 | 868 | 81 | ||
| AK217962.1 | - | 1 - 2 | 62 | 81 | ||
| BC016080.1 | - | 4 - 5 | 871 | 4489287 | 81 | |
| BC085483.1 | - | 4 - 5 | 850 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpn1
Genomic Map