Assay Details
| Gene Symbol: Rpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133933.4 | NP_598694.3 | 5 - 6 | 1099 | 66 | |
| GenBank mRNA | AK029923.1 | - | 5 - 6 | 1057 | 66 | |
| AK052130.1 | - | 5 - 6 | 1053 | 66 | ||
| AK132750.1 | - | 5 - 6 | 1072 | 66 | ||
| AK151899.1 | - | 5 - 6 | 1063 | 66 | ||
| AK152003.1 | - | 5 - 6 | 1072 | 66 | ||
| AK153867.1 | - | 5 - 6 | 1058 | 66 | ||
| AK217962.1 | - | 2 - 3 | 252 | 66 | ||
| BC002175.1 | - | 1 - 2 | 58 | 66 | ||
| BC016080.1 | - | 5 - 6 | 1061 | 4489287 | 66 | |
| BC085483.1 | - | 5 - 6 | 1040 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpn1
Genomic Map