Assay Details
| Gene Symbol: Isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133934.4 | NP_598695.1 | 5 - 6 | 305 | 107 | |
| GenBank mRNA | AK173116.1 | - | 273 | 107 | ||
| BC020103.1 | - | 5 - 6 | 269 | 3673446 | 107 | |
| BC037695.1 | - | 5 - 6 | 310 | 4009615 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Isy1
Genomic Map