Assay Details
| Gene Symbol: Unc45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133952.2 | NP_598713.2 | 15 - 16 | 2170 | 79 | |
| GenBank mRNA | AK028912.1 | - | 15 - 16 | 2156 | 79 | |
| AK047292.1 | - | 15 - 16 | 2117 | 79 | ||
| AK047476.1 | - | 15 - 16 | 2117 | 79 | ||
| AK088034.1 | - | 15 - 16 | 2091 | 79 | ||
| AK166809.1 | - | 15 - 16 | 2153 | 79 | ||
| BC004717.1 | - | 15 - 16 | 2159 | 3582116 | 79 | |
More Information
Set Membership: |
|

Unc45a
Genomic Map