Assay Details
| Gene Symbol: Usb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133954.2 | NP_598715.2 | 2 - 3 | 350 | 58 | |
| GenBank mRNA | AK135527.1 | - | 2 - 3 | 343 | 58 | |
| AK160901.1 | - | 2 - 3 | 333 | 58 | ||
| AK168683.1 | - | 2 - 3 | 332 | 58 | ||
| BC016418.1 | - | 2 - 3 | 294 | 4224590 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usb1
Genomic Map