Assay Details
Gene Symbol: Mbtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134012.3 | NP_598773.2 | 7 - 8 | 986 | 103 | |
GenBank mRNA | AK033238.1 | - | 2 - 3 | 3859 | 103 | |
AK141155.1 | - | 7 - 8 | 981 | 103 | ||
AK153919.1 | - | 9 - 10 | 821 | 103 | ||
BC020018.1 | - | 3 - 4 | 297 | 2646754 | 103 | |
BC062907.1 | - | 7 - 8 | 986 | 6813849 | 103 | |
BC064014.1 | - | 9 - 10 | 1090 | 6335954 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
