Assay Details
| Gene Symbol: Aldh6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134042.2 | NP_598803.1 | 5 - 6 | 438 | 65 | |
| GenBank mRNA | AF297860.1 | - | 5 - 6 | 438 | 65 | |
| AK033587.1 | - | 5 - 6 | 465 | 65 | ||
| AK082790.1 | - | 5 - 6 | 492 | 65 | ||
| AK128924.1 | - | 5 - 6 | 470 | 65 | ||
| AK147146.1 | - | 5 - 6 | 492 | 65 | ||
| AK155814.1 | - | 5 - 6 | 489 | 65 | ||
| AK164366.1 | - | 4 - 5 | 2038 | 65 | ||
| AK169915.1 | - | 5 - 6 | 478 | 65 | ||
| AK170305.1 | - | 5 - 6 | 491 | 65 | ||
| AK171581.1 | - | 5 - 6 | 492 | 65 | ||
| AK171896.1 | - | 5 - 6 | 481 | 65 | ||
| AK212397.1 | - | 1 - 2 | 52 | 65 | ||
| AK217579.1 | - | 1 - 2 | 39 | 65 | ||
| AK219365.1 | - | 1 - 2 | 59 | 65 | ||
| BC031148.1 | - | 5 - 6 | 563 | 4975560 | 65 | |
| BC115855.1 | - | 5 - 6 | 413 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh6a1
Genomic Map