Assay Details
| Gene Symbol: Adi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134052.2 | NP_598813.1 | 3 - 4 | 463 | 102 | |
| GenBank mRNA | AK089966.1 | - | 3 - 4 | 447 | 102 | |
| AK145831.1 | - | 3 - 4 | 440 | 102 | ||
| BC005695.1 | - | 3 - 4 | 463 | 3597652 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular amino acid biosynthetic process
methionine biosynthetic process
L-methionine biosynthetic process from methylthioadenosine
oxidation-reduction process
acireductone dioxygenase [iron(II)-requiring] activity
oxidoreductase activity
metal ion binding
dioxygenase activity
cellular amino acid biosynthetic process
methionine biosynthetic process
L-methionine biosynthetic process from methylthioadenosine
oxidation-reduction process
acireductone dioxygenase [iron(II)-requiring] activity
oxidoreductase activity
metal ion binding
dioxygenase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28314427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28314431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28314428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28314430 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121913 | BC005695.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11773 | AK145831.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11773 | NM_134052.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565197 | AK038419.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307615 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103089 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121913 | BC005695.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11773 | AK145831.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11773 | NM_134052.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P224263 | BC005695.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28314425 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103089 | - | - |
| Whole Mouse Genome G4122A | A_51_P224263 | BC005695.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140174 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103089 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10317799 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10006270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28314428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28314431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28314427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28314430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28314405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10317798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09856451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10318563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10414572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10405157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09856453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09812456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10051709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09758117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09856452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10413592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10016834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10006270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09826774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09856449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09812458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09793061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10413591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10386305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10318558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09764851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28314401 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395063 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438759_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438758_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449076_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395058 | - | - |
| Mouse Genome 430 2.0 Array | 1438759_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438759_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438758_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438758_at | - | - |
| Mouse Genome 430 2.0 Array | 1449076_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1449076_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 104923_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 104923_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200001822.mm.1 | NM_134052.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001822.mm.1 | NM_134052.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001822.mm.1 | TC1200000236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200001825.mm.1 | NM_134052.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001825.mm.1 | NM_134052.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001825.mm.1 | TC1200000236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200001821.mm.1 | NM_134052.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001821.mm.1 | NM_134052.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001821.mm.1 | TC1200000236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200001820.mm.1 | NM_134052.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001820.mm.1 | NM_134052.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001820.mm.1 | TC1200000236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200001818.mm.1 | NM_134052.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001818.mm.1 | NM_134052.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001818.mm.1 | TC1200000236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200001824.mm.1 | NM_134052.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001824.mm.1 | NM_134052.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001824.mm.1 | TC1200000236.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438759_x_at | - | - |
| Mouse Expression Set 430 | 1438759_PM_x_at | - | - |
| Mouse Expression Set 430 | 1449076_x_at | - | - |
| Mouse Expression Set 430 | 1438758_at | - | - |
| Mouse Expression Set 430 | 1449076_PM_x_at | - | - |
| Mouse Expression Set 430 | 1438758_PM_at | - | - |
| Murine Genome U74v2 Set | 103429_PM_i_at | - | - |
| Murine Genome U74v2 Set | 103429_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449076_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438759_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438758_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449076_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438758_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438759_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438759_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449076_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449076_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438758_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438759_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438758_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 520635 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6220546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006220546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231567 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231567 | - | - |
| mouse-6 v1.1 (Illumina) | scl0104923.5_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6220546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0104923.5_14-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6220546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3065603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231567 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3143557 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3143557 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231567 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3065603 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0104923.5_14-S | - | - |

Adi1
Genomic Map