Assay Details
| Gene Symbol: Slc35b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170430.1 | NP_001163901.1 | 4 - 5 | 694 | 81 | |
| NM_001170431.1 | NP_001163902.1 | 4 - 5 | 829 | 81 | ||
| NM_134060.4 | NP_598821.1 | 4 - 5 | 672 | 81 | ||
| GenBank mRNA | AK076592.1 | - | 4 - 5 | 667 | 81 | |
| AK077208.1 | - | 4 - 5 | 648 | 81 | ||
| AK141280.1 | - | 4 - 5 | 830 | 81 | ||
| BC006881.1 | - | 4 - 5 | 686 | 3596363 | 81 | |
| CX238927.1 | - | 3 - 4 | 378 | 81 | ||
More Information
Set Membership: |
|

Slc35b3
Genomic Map