Assay Details
Gene Symbol: Tomm70a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138599.5 | NP_613065.2 | 6 - 7 | 1252 | 88 | |
GenBank mRNA | AK012084.1 | - | 6 - 7 | 1253 | 88 | |
AK083586.1 | - | 6 - 7 | 1253 | 88 | ||
AK122356.1 | - | 1185 | 88 | |||
AK145458.1 | - | 6 - 7 | 1265 | 88 | ||
AK150429.1 | - | 6 - 7 | 1253 | 88 | ||
AK161720.1 | - | 6 - 7 | 1242 | 88 | ||
AK169809.1 | - | 6 - 7 | 1214 | 88 | ||
BC057096.1 | - | 6 - 7 | 1253 | 5709110 | 88 | |
BC139420.1 | - | 6 - 7 | 1225 | 8862442 | 88 | |
BC139421.1 | - | 6 - 7 | 1225 | 8862443 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
