Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400010847.mm.1 |
NM_138683.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010847.mm.1 |
NM_138683.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010847.mm.1 |
TC0400001433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010842.mm.1 |
NM_138683.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010842.mm.1 |
NM_138683.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010842.mm.1 |
TC0400001433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010844.mm.1 |
NM_138683.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010844.mm.1 |
NM_138683.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010844.mm.1 |
TC0400001433.mm.1 |
MTA TC |