Assay Details
Gene Symbol: Spag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139151.4 | NP_631890.3 | 8 - 9 | 946 | 70 | |
GenBank mRNA | AB041554.1 | - | 6 - 7 | 1525 | 70 | |
AK028080.1 | - | 8 - 9 | 915 | 70 | ||
AK043812.1 | - | 8 - 9 | 914 | 70 | ||
AK220494.1 | - | 2219 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
