Assay Details
| Gene Symbol: Spag4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139151.4 | NP_631890.3 | 9 - 10 | 1064 | 62 | |
| GenBank mRNA | AB041554.1 | - | 7 - 8 | 1643 | 62 | |
| AK028080.1 | - | 9 - 10 | 1033 | 62 | ||
| AK220494.1 | - | 2337 | 62 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Spag4
Genomic Map