Assay Details
| Gene Symbol: Plac8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139198.2 | NP_631937.1 | 1 - 2 | 53 | 65 | |
| GenBank mRNA | AJ422151.1 | - | 1 - 2 | 73 | 65 | |
| AK007369.1 | - | 1 - 2 | 54 | 65 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ422151.1, AK007369.1, NM_139198.2 transcripts.
Set Membership: |
|

Plac8
Genomic Map