Assay Details
| Gene Symbol: Cdkal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082629.1 | - | 10 - 11 | 1033 | 75 | |
| AK158546.1 | - | 10 - 11 | 1023 | 75 | ||
| AK169761.1 | - | 10 - 11 | 1030 | 75 | ||
| BC016073.1 | - | 10 - 11 | 974 | 4482130 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdkal1
Genomic Map