Assay Details
Gene Symbol: Lgi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145219.4 | NP_660254.1 | 6 - 7 | 907 | 68 | |
GenBank mRNA | AF515590.1 | - | 6 - 7 | 889 | 68 | |
AK047495.1 | - | 6 - 7 | 859 | 68 | ||
AK049831.1 | - | 6 - 7 | 859 | 68 | ||
AK087125.1 | - | 7 - 8 | 1046 | 68 | ||
AK132296.1 | - | 6 - 7 | 901 | 68 | ||
AY174077.1 | - | 6 - 7 | 889 | 68 | ||
BC055315.1 | - | 6 - 7 | 767 | 5686778 | 68 | |
BC061460.1 | - | 6 - 7 | 767 | 5686778 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
