Assay Details
| Gene Symbol: Dhx35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291144.1 | NP_001278073.1 | 5 - 6 | 549 | 77 | |
| NM_145742.2 | NP_665685.1 | 5 - 6 | 529 | 77 | ||
| GenBank mRNA | AK004665.1 | - | 5 - 6 | 475 | 77 | |
| AK155482.1 | - | 5 - 6 | 468 | 77 | ||
| AK156955.1 | - | 5 - 6 | 495 | 77 | ||
| AK170952.1 | - | 5 - 6 | 468 | 77 | ||
| BC029709.1 | - | 5 - 6 | 516 | 5359253 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291144.1 transcripts.
Set Membership: |
|

Dhx35
Genomic Map