Assay Details
| Gene Symbol: Aars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146217.4 | NP_666329.2 | 16 - 17 | 2724 | 81 | |
| GenBank mRNA | AK044451.1 | - | 16 - 17 | 2475 | 81 | |
| AK085725.1 | - | 16 - 17 | 2665 | 81 | ||
| AK150227.1 | - | 16 - 17 | 2652 | 81 | ||
| AK158139.1 | - | 16 - 17 | 2673 | 81 | ||
| AY223875.1 | - | 16 - 17 | 2664 | 81 | ||
| BC013261.1 | - | 13 - 14 | 1949 | 81 | ||
| BC026611.1 | - | 5 - 6 | 753 | 81 | ||
| BC033273.1 | - | 2599 | 4196789 | 81 | ||
| BC058620.1 | - | 6 - 7 | 898 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aars
Genomic Map