Assay Details
| Gene Symbol: Adcy8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291903.1 | NP_001278832.1 | 11 - 12 | 3772 | 87 | |
| NM_009623.2 | NP_033753.2 | 12 - 13 | 3862 | 87 | ||
| GenBank mRNA | AK138244.1 | - | 11 - 12 | 3773 | 87 | |
| BC059010.1 | - | 12 - 13 | 2959 | 87 | ||
| BC094231.1 | - | 12 - 13 | 3300 | 87 | ||
| U85021.1 | - | 12 - 13 | 3722 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcy8
Genomic Map